siRNA Details
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138547.2 | NP_612556.1 | 8 | 952 | ||
GenBank mRNA | BC091123.1 | - | 8 | 952 | 7372929 | |
D17310.1 | - | 8 | 1140 | |||
M64393.1 | - | 8 | 951 | |||
S57790.1 | - | 8 | 910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
