siRNA Details
Gene Symbol: Dab2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138710.3 | NP_619724.3 | 6 | 893 | ||
XM_006234057.2 | XP_006234119.1 | 5 | 880 | |||
XM_008761697.1 | XP_008759919.1 | 3 | 1291 | |||
XM_008761698.1 | XP_008759920.1 | 3 | 1291 | |||
XM_008761699.1 | XP_008759921.1 | 3 | 1292 | |||
GenBank mRNA | AF236130.1 | - | 5 | 601 | ||
BC061865.1 | - | 2 | 207 | 5598046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
