siRNA Details
Gene Symbol: Hip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100475.1 | NP_001093945.1 | 28 | 2909 | ||
XM_006249098.2 | XP_006249160.1 | 28 | 3021 | |||
XM_006249099.2 | XP_006249161.1 | 28 | 3059 | |||
XM_006249100.2 | XP_006249162.1 | 28 | 2951 | |||
XM_008769114.1 | XP_008767336.1 | 28 | 3021 | |||
GenBank mRNA | AF388529.1 | - | 5 | 445 |
