siRNA Details
Gene Symbol: Sftpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138842.1 | NP_620197.1 | 4 | 299 | ||
XM_006236657.2 | XP_006236719.1 | 4 | 334 | |||
GenBank mRNA | BC072466.1 | - | 4 | 298 | 7100971 | |
X14778.1 | - | 4 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
