siRNA Details
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138848.1 | NP_620203.1 | 5, 6 | 1170 | ||
XM_008762758.1 | XP_008760980.1 | 5, 6 | 1330 | |||
GenBank mRNA | AB020726.1 | - | 5, 6 | 1243 | ||
AF109393.1 | - | 5, 6 | 1170 | |||
BC070886.1 | - | 5, 6 | 1240 | 7098164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
