siRNA Details
Gene Symbol: Syt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138849.1 | NP_620204.1 | 7 | 1248 | ||
XM_006230101.2 | XP_006230163.1 | 7 | 1343 | |||
XM_006230102.2 | XP_006230164.1 | 7 | 1228 | |||
XM_006230104.1 | XP_006230166.1 | 7 | 1442 | |||
XM_008759675.1 | XP_008757897.1 | 5 | 1708 | |||
GenBank mRNA | U30831.1 | - | 7 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
