siRNA Details
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138853.2 | NP_620208.1 | 3, 4 | 166 | ||
XM_006239000.1 | XP_006239062.1 | 3, 4 | 228 | |||
XM_006239002.1 | XP_006239064.1 | 3, 4 | 228 | |||
XM_008764131.1 | XP_008762353.1 | 1, 2 | 29 | |||
GenBank mRNA | AB072920.1 | - | 2, 3 | 94 | ||
BC078730.1 | - | 3, 4 | 166 | 7134156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
