siRNA Details
Gene Symbol: Slc38a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138854.1 | NP_620209.1 | 14 | 1314 | ||
XM_008773068.1 | XP_008771290.1 | 14 | 1246 | |||
GenBank mRNA | AF276870.1 | - | 14 | 1314 | ||
BC128725.1 | - | 14 | 1246 | 8362014 |
