siRNA Details
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138856.2 | NP_620211.2 | 4 | 432 | ||
XM_008760746.1 | XP_008758968.1 | 8 | 1308 | |||
GenBank mRNA | AF492384.1 | - | 3 | 355 | ||
BC086587.1 | - | 4 | 432 | |||
BC158554.1 | - | 4 | 651 | 7380100 |
