siRNA Details
Gene Symbol: Rasgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105753.1 | NP_001099223.1 | 1 | 309 | ||
NM_001170531.1 | NP_001164002.1 | 1 | 233 | |||
GenBank mRNA | CB607130.1 | - | 1 | 108 | ||
CB713048.1 | - | 1 | 245 | |||
X67241.1 | - | 1 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
