siRNA Details
Gene Symbol: Rasgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170531.1 | NP_001164002.1 | 18 | 2720 | ||
GenBank mRNA | BC091317.1 | - | 10 | 1370 | ||
X67241.1 | - | 18 | 2659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
