siRNA Details
Gene Symbol: Atp5o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138883.1 | NP_620238.1 | 2 | 78 | ||
GenBank mRNA | BC060544.1 | - | 2 | 86 | 6918775 | |
D13127.1 | - | 2 | 78 | |||
FQ209940.1 | - | 2 | 86 | |||
FQ210807.1 | - | 2 | 92 | |||
FQ217675.1 | - | 2 | 83 | |||
FQ223905.1 | - | 2 | 115 | |||
FQ226488.1 | - | 2 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
