siRNA Details
Gene Symbol: Atp5o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138883.1 | NP_620238.1 | 7 | 570 | ||
GenBank mRNA | BC060544.1 | - | 7 | 578 | 6918775 | |
D13127.1 | - | 7 | 570 | |||
FQ209940.1 | - | 7 | 578 | |||
FQ210807.1 | - | 7 | 584 | |||
FQ217675.1 | - | 7 | 575 | |||
FQ223905.1 | - | 7 | 607 | |||
FQ226488.1 | - | 7 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
