siRNA Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138884.1 | NP_620239.1 | 3 | 384 | ||
XM_006236285.1 | XP_006236347.1 | 3 | 404 | |||
GenBank mRNA | D17309.1 | - | 3 | 384 | ||
FQ211413.1 | - | 3 | 402 | |||
FQ219185.1 | - | 3 | 403 | |||
FQ219730.1 | - | 3 | 347 | |||
S80431.1 | - | 3 | 356 |
