siRNA Details
Gene Symbol: Hspb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138887.1 | NP_620242.1 | 3 | 1035 | ||
GenBank mRNA | D29960.1 | - | 3 | 1035 | ||
FQ214730.1 | - | 3 | 1060 | |||
FQ215336.1 | - | 3 | 1064 | |||
FQ217608.1 | - | 3 | 1065 | |||
FQ224974.1 | - | 3 | 1066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
