siRNA Details
Gene Symbol: Phf5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138888.1 | NP_620243.1 | 3 | 233 | ||
GenBank mRNA | AF495522.1 | - | 3 | 233 | ||
AY321339.1 | - | 249 | ||||
BC168144.1 | - | 3 | 251 | 8372591 |

More Information
Other Species Targeted

Gene Ontology Categories:
