siRNA Details
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138892.1 | NP_620247.1 | 3 | 405 | ||
GenBank mRNA | AF331839.1 | - | 3 | 379 | ||
AY029335.1 | - | 3 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
