siRNA Details
Gene Symbol: Ppp1r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138912.2 | NP_620267.2 | 2 | 473 | ||
XM_006253215.2 | XP_006253277.1 | 2 | 1039 | |||
XM_006253216.2 | XP_006253278.1 | 2 | 360 | |||
GenBank mRNA | BC072514.1 | - | 2 | 473 | 7104413 | |
Y18208.1 | - | 2 | 336 |

More Information
Other Species Targeted

Gene Ontology Categories:
