siRNA Details
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138914.1 | NP_620269.1 | 1705 | |||
GenBank mRNA | AB073208.1 | - | 1705 | |||
AB126168.1 | - | 1618 | ||||
AB126169.1 | - | 1522 | ||||
AB126170.1 | - | 1690 | ||||
AB126171.1 | - | 1603 | ||||
AB126172.1 | - | 1507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
