siRNA Details
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138914.1 | NP_620269.1 | 1700 | |||
GenBank mRNA | AB073208.1 | - | 1700 | |||
AB126168.1 | - | 1613 | ||||
AB126169.1 | - | 1517 | ||||
AB126170.1 | - | 1685 | ||||
AB126171.1 | - | 1598 | ||||
AB126172.1 | - | 1502 |
