siRNA Details
Gene Symbol: Caly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190399.1 | NP_001177328.1 | 4 | 367 | ||
NM_138915.1 | NP_620270.1 | 4 | 426 | |||
GenBank mRNA | AF303658.1 | - | 4 | 426 | ||
FQ213422.1 | - | 4 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
