siRNA Details
Gene Symbol: Prokr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138977.1 | NP_620433.1 | 1 | 448 | ||
XM_006236821.2 | XP_006236883.1 | 2 | 743 | |||
GenBank mRNA | AB158419.1 | - | 1 | 448 | ||
AB158420.1 | - | 1 | 448 | |||
AY089974.1 | - | 1 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
