siRNA Details
Gene Symbol: Prokr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138978.1 | NP_620434.1 | 2 | 1114 | ||
XM_006235059.2 | XP_006235121.1 | 4 | 1523 | |||
GenBank mRNA | AY089975.1 | - | 2 | 1114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
