siRNA Details
Gene Symbol: Dnah8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772852.1 | XP_008771074.1 | 12 | 1924 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnah8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772852.1 | XP_008771074.1 | 12 | 1924 |