siRNA Details
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181363.2 | NP_852028.1 | 3, 4 | 518 | ||
XM_008760905.1 | XP_008759127.1 | 3, 4 | 325 | |||
GenBank mRNA | AF188995.1 | - | 2, 3 | 293 | ||
U58289.1 | - | 3, 4 | 518 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153561, AF453427, AF453428, BC146000, AK145299, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
