siRNA Details
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181363.2 | NP_852028.1 | 2, 3 | 474 | ||
XM_008760905.1 | XP_008759127.1 | 2, 3 | 281 | |||
GenBank mRNA | U58289.1 | - | 2, 3 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
