siRNA Details
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768788.1 | XP_008767010.1 | 6 | 1029 | ||
GenBank mRNA | U89744.1 | - | 5 | 991 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768788.1 | XP_008767010.1 | 6 | 1029 | ||
GenBank mRNA | U89744.1 | - | 5 | 991 |