siRNA Details
Gene Symbol: Exoc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1629 | ||
GenBank mRNA | AF032669.1 | - | 1 | 1629 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198103, BC057052.
Gene Ontology Categories:
