siRNA Details
Gene Symbol: Impg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139088.2 | NP_620788.2 | 11 | 1219 | ||
XM_008768628.1 | XP_008766850.1 | 11 | 2207 | |||
XM_008768629.1 | XP_008766851.1 | 6 | 567 | |||
GenBank mRNA | U76717.1 | - | 11 | 1219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
