siRNA Details
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139089.1 | NP_620789.1 | 2 | 246 | ||
GenBank mRNA | BC058444.1 | - | 2 | 261 | 6918440 | |
FQ225009.1 | - | 2 | 260 | |||
FQ225217.1 | - | 2 | 260 | |||
FQ225221.1 | - | 2 | 260 | |||
FQ225582.1 | - | 2 | 257 | |||
FQ225670.1 | - | 2 | 260 | |||
FQ226479.1 | - | 2 | 257 | |||
FQ231406.1 | - | 2 | 260 | |||
FQ232883.1 | - | 2 | 260 | |||
FQ233441.1 | - | 2 | 260 | |||
FQ233623.1 | - | 2 | 260 | |||
FQ233884.1 | - | 2 | 260 | |||
U17035.1 | - | 2 | 246 | |||
U22520.1 | - | 2 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
