siRNA Details
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139098.1 | NP_620798.1 | 15, 16 | 2110 | ||
XM_006253601.2 | XP_006253663.1 | 15, 16 | 2140 | |||
GenBank mRNA | BC107590.1 | - | 15, 16 | 1976 | 7461248 | |
U25746.1 | - | 15, 16 | 2110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
