siRNA Details
Gene Symbol: Atp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139099.1 | NP_620799.1 | 2 | 148 | ||
GenBank mRNA | AF010323.1 | - | 2 | 148 | ||
BC058133.1 | - | 2 | 138 | 6887013 | ||
FQ211630.1 | - | 2 | 170 | |||
FQ211898.1 | - | 2 | 182 | |||
FQ214112.1 | - | 2 | 170 | |||
FQ214935.1 | - | 2 | 173 | |||
FQ215084.1 | - | 2 | 100 | |||
FQ216950.1 | - | 2 | 170 | |||
FQ217027.1 | - | 2 | 134 | |||
FQ217160.1 | - | 2 | 124 | |||
FQ217179.1 | - | 2 | 173 | |||
FQ217559.1 | - | 2 | 123 | |||
FQ217635.1 | - | 2 | 129 | |||
FQ218023.1 | - | 2 | 129 | |||
FQ218046.1 | - | 2 | 129 | |||
FQ222358.1 | - | 2 | 157 | |||
FQ223590.1 | - | 2 | 182 | |||
FQ223710.1 | - | 2 | 129 | |||
FQ223763.1 | - | 2 | 170 | |||
FQ223780.1 | - | 2 | 158 | |||
FQ223794.1 | - | 2 | 175 | |||
FQ223992.1 | - | 2 | 152 | |||
FQ224060.1 | - | 2 | 129 | |||
FQ224189.1 | - | 2 | 173 | |||
FQ224205.1 | - | 2 | 173 | |||
FQ224455.1 | - | 2 | 172 | |||
FQ224486.1 | - | 2 | 129 | |||
FQ224537.1 | - | 2 | 173 | |||
FQ224608.1 | - | 2 | 170 | |||
FQ224652.1 | - | 2 | 134 | |||
FQ224692.1 | - | 2 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
