siRNA Details
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139106.1 | NP_620806.1 | 2, 3 | 337 | ||
XM_006240845.2 | XP_006240907.1 | 3, 4 | 852 | |||
GenBank mRNA | FQ214102.1 | - | 545 | |||
FQ223899.1 | - | 2, 3 | 385 | |||
U00926.1 | - | 2, 3 | 337 |
