siRNA Details
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139106.1 | NP_620806.1 | 2 | 333 | ||
XM_006240845.2 | XP_006240907.1 | 3 | 848 | |||
GenBank mRNA | BC161836.1 | - | 3 | 858 | 9098303 | |
FQ214102.1 | - | 541 | ||||
FQ223899.1 | - | 2 | 381 | |||
U00926.1 | - | 2 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
