siRNA Details
Gene Symbol: Commd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139108.2 | NP_620808.1 | 493 | |||
XM_006241924.2 | XP_006241986.1 | 2 | 806 | |||
XM_006241925.2 | XP_006241987.1 | 2 | 796 | |||
XM_006241926.2 | XP_006241988.1 | 2 | 784 | |||
XM_008765544.1 | XP_008763766.1 | 2 | 827 | |||
GenBank mRNA | AF290194.1 | - | 498 |
Gene Symbol: LOC100910472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241915.2 | XP_006241977.1 | 2 | 806 | ||
XM_006241916.2 | XP_006241978.1 | 2 | 822 | |||
XM_006241917.2 | XP_006241979.1 | 2 | 796 | |||
XM_006241918.2 | XP_006241980.1 | 2 | 784 | |||
XM_008765634.1 | XP_008763856.1 | 2 | 827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
