siRNA Details
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139111.1 | NP_620811.1 | 3 | 434 | ||
GenBank mRNA | AF253064.2 | - | 2 | 275 | ||
AF253065.2 | - | 3 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
