siRNA Details
Gene Symbol: Nr2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139113.1 | NP_620813.1 | 1, 2 | 500 | ||
GenBank mRNA | AF003926.1 | - | 1, 2 | 500 |

More Information
Other Species Targeted

Gene Ontology Categories:
