siRNA Details
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139115.2 | NP_620815.2 | 8, 9 | 1079 | ||
XM_006246885.2 | XP_006246947.1 | 9, 10 | 2156 | |||
XM_006246886.2 | XP_006246948.1 | 9, 10 | 2158 | |||
XM_006246887.2 | XP_006246949.1 | 9, 10 | 2156 | |||
XM_006246888.2 | XP_006246950.1 | 9, 10 | 2158 | |||
XM_006246889.2 | XP_006246951.1 | 8, 9 | 2036 | |||
XM_006246890.2 | XP_006246952.1 | 8, 9 | 2036 | |||
GenBank mRNA | AF140359.1 | - | 9, 10 | 1452 | ||
BC080241.1 | - | 8, 9 | 2618 | |||
BC098656.1 | - | 8, 9 | 1079 | 7192854 |
