siRNA Details
Gene Symbol: Gng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139185.1 | NP_631924.1 | 3 | 266 | ||
XM_006228384.2 | XP_006228446.1 | 2 | 620 | |||
XM_006228385.2 | XP_006228447.1 | 2 | 64 | |||
GenBank mRNA | L35921.1 | - | 3 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
