siRNA Details
Gene Symbol: Klrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139187.2 | NP_631926.1 | 5, 6 | 510 | ||
GenBank mRNA | AF416564.2 | - | 5, 6 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
