siRNA Details
Gene Symbol: Ppap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139252.1 | NP_640345.1 | 2 | 183 | ||
XM_008765155.1 | XP_008763377.1 | 2 | 166 | |||
GenBank mRNA | AF503611.1 | - | 2 | 183 | ||
BC062088.1 | - | 2 | 139 | 5621318 |
