siRNA Details
Gene Symbol: Ly6i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139257.1 | NP_640350.1 | 4 | 368 | ||
XM_006241765.2 | XP_006241827.1 | 355 | ||||
XM_006241766.2 | XP_006241828.1 | 5 | 495 | |||
XM_008765545.1 | XP_008763767.1 | 5 | 482 | |||
GenBank mRNA | M30689.1 | - | 4 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
