siRNA Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248989.2 | XP_006249051.1 | 7 | 3256 | ||
GenBank mRNA | AF030243.1 | - | 2 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
