siRNA Details
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139216.1 | NP_631962.1 | 2 | 1280 | ||
NM_139217.1 | NP_631963.1 | 2 | 1237 | |||
XM_006241327.2 | XP_006241389.1 | 3 | 1489 | |||
GenBank mRNA | M34052.1 | - | 2 | 1280 | ||
M59211.1 | - | 2 | 1237 | |||
M84202.1 | - | 3 | 1294 | |||
M84203.1 | - | 2 | 1280 | |||
X62839.1 | - | 3 | 1391 |

More Information
Other Species Targeted

Gene Ontology Categories:
