siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139263.1 | NP_640356.1 | 8 | 2344 | ||
XM_008768850.1 | XP_008767072.1 | 8 | 1720 | |||
GenBank mRNA | AF038388.1 | - | 8 | 2344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
