siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139263.1 | NP_640356.1 | 5, 6 | 1968 | ||
XM_008768850.1 | XP_008767072.1 | 5, 6 | 1344 | |||
GenBank mRNA | AF038388.1 | - | 5, 6 | 1968 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_139233, NM_139232, NM_139234, BC129903, AF402613, and others.
Gene Ontology Categories:
