siRNA Details
Gene Symbol: Dus1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172018.1 | NP_742015.1 | 2 | 274 | ||
XM_006247874.1 | XP_006247936.1 | 2 | 467 | |||
XM_006247875.1 | XP_006247937.1 | 2 | 467 | |||
GenBank mRNA | AF508021.1 | - | 2 | 274 | ||
AY327509.1 | - | 2 | 274 | |||
AY327510.1 | - | 2 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
