siRNA Details
Gene Symbol: Ifi47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172019.2 | NP_742016.2 | 2 | 977 | ||
XM_006246175.2 | XP_006246237.1 | 2 | 2009 | |||
XM_008767662.1 | XP_008765884.1 | 2 | 1197 | |||
GenBank mRNA | AF508023.1 | - | 2 | 995 | ||
BC066660.1 | - | 2 | 948 | 5623225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
