siRNA Details
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198050.1 | NP_932167.1 | 15, 16 | 3507 | ||
XM_006253556.2 | XP_006253618.1 | 15, 16 | 3651 | |||
XM_006253557.2 | XP_006253619.1 | 14, 15 | 3507 | |||
XM_008771424.1 | XP_008769646.1 | 15, 16 | 3675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
