siRNA Details
Gene Symbol: Sipa1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139330.1 | NP_647546.1 | 2 | 1615 | ||
GenBank mRNA | AF026504.1 | - | 2 | 1615 | ||
AY043226.1 | - | 1 | 1178 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sipa1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139330.1 | NP_647546.1 | 2 | 1615 | ||
GenBank mRNA | AF026504.1 | - | 2 | 1615 | ||
AY043226.1 | - | 1 | 1178 |